Skip to Content
Merck
All Photos(1)

Key Documents

940186

Sigma-Aldrich

LiBETI

new

electrolyte powder, anhydrous, battery grade

Synonym(s):

Bis(pentafluoroethanesulfonyl)imide lithium salt, LiBETA, Lithium bis(pentafluoroethylsulfonyl)imide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4 HF10 NO4 S2Li
CAS Number:
Molecular Weight:
388.11
UNSPSC Code:
12352107

grade

battery grade

Quality Level

Assay

99%

form

powder

mp

328 °C

application(s)

battery manufacturing
battery manufacturing

SMILES string

[Li].O=S(=O)(NS(=O)(=O)C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)F

InChI

1S/C4HF10NO4S2.Li/c5-1(6,7)3(11,12)20(16,17)15-21(18,19)4(13,14)2(8,9)10;/h15H;

InChI key

TWWUUDFJDGCFDD-UHFFFAOYSA-N

General description

Lithium bis(pentafluoroethylsulfonyl)imide (LiBETI) is an electrolytic salt used in lithium-ion electrolytes. It is a white powder with a high melting point of 328 °C that is soluble in many polar solvents including water, acetonitrile, and many of the carbonates used in traditional lithium-ion liquid electrolytes.

Application

LiBETI finds use in lithium-ion batteries, including organic liquid electrolytes, water-in-salt electrolytes, and polymer electrolytes. LiBETI has been found to have much better durability against moisture than LiPF6 or LiBF4, especially at high operating temperatures. Additionally, LiBETI is less corrosive to aluminum current collectors than other imide-based electrolytic salts like LiTFSI.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service