Skip to Content
Merck
All Photos(1)

Key Documents

939781

t-BuSF

new

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H23FN2O2S
CAS Number:
Molecular Weight:
266.38
UNSPSC Code:
12352142

Quality Level

Assay

≥95%

form

powder or crystals

color

white to yellow

storage temp.

2-8°C

SMILES string

CC(C)(C)S(=NC(N(C(C)C)C(C)C)=O)(F)=O

InChI

InChI=1S/C11H23FN2O2S/c1-8(2)14(9(3)4)10(15)13-17(12,16)11(5,6)7/h8-9H,1-7H3/t17-/m0/s1

InChI key

ZDAOBEFNHRJWST-KRWDZBQOSA-N

General description

t-BuSF is a novel chiral sulfur fluorine exchange (SuFEx) reagent. SuFEx type click chemistry is an effective method to synthesize many molecules of interest to medicinal chemists, chemical biologists, or material scientists. While widely utilized, methods to use SuFEx chemistry chirally are limited. t-BuSF provides a method to rapidly synthesize asymmetric sulfoximine, sulfonimidoyl flourides and sulfonimidamides with high yield and enantiomeric excess.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service