Skip to Content
Merck
All Photos(1)

Key Documents

936642

Sigma-Aldrich

9-Bromoanthracene-d9

≥98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12142200

isotopic purity

≥98 atom % D

Quality Level

Assay

≥97% (CP)

form

liquid
semisolid
solid

packaging

pkg of 1 g
pkg of 10 g

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service