Skip to Content
Merck
All Photos(3)

Key Documents

936219

Sigma-Aldrich

AdQPhos Pd(crotyl)Cl

greener alternative
Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C64H66ClFePPd
Molecular Weight:
1063.90
UNSPSC Code:
12352142
NACRES:
NA.21

form

powder

Quality Level

reaction suitability

reaction type: Cross Couplings

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

light red to red, and Pink

functional group

(palladium)
phosphine

greener alternative category

SMILES string

Cl[Pd]([P](C12C[C@H]3C[C@H](C[C@H](C3)C2)C1)(C45C[C@H]6C[C@H](C[C@H](C6)C5)C4)c7cccc7)C.CCCC.c8(C9=CC=CC=C9)c(C%10=CC=CC=C%10)c(C%11=CC=CC=C%11)c(C%12=CC=CC=C%12)c8C%13=CC=CC=C%13.[Fe]

General description

The precatalysts derived from AdQPhos exhibit excellent activity towards palladium-catalyzed α-arylation reactions under milder conditions. The strength of these catalytic systems in enabling such reactions were demonstrated with the broad scope of nucleophiles, including nitroalkanes, nitriles, amides, esters, and ketones. Compared to other state-of-the-art catalysts, this system was far superior and served as the first versatile catalytic system to accommodate all these nucleophiles in this context.

Further, by merging with micellar catalysis, this catalytic system demonstrated the feasibility of enabling α-arylation reactions under mild aqueous conditions. This approach provides an environmentally benign alternative to toxic/hazardous solvents like 1,4-dioxane or NMP, bringing modern organic synthesis closer to being ideally sustainable.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

In addition to facilitating α-arylations, these catalysts demonstrate compatibility with various challenging cross-coupling reactions, such as Suzuki-Miyaura, Buchwald-Hartwig aminations, and C-O and C-S cross-couplings.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service