Skip to Content
Merck
All Photos(1)

Key Documents

936049

Sigma-Aldrich

Sodium bis(trifluoromethylsulfonyl)imide

new
greener alternative

≥99.5% trace metals basis, anhydrous, battery grade

Synonym(s):

Sodium trifluoromethanesulfonimide, NaTFSI, Sodium triflimide, Sodium bis(trifluoromethylsulfonyl)imide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2F6NNaO4S2
CAS Number:
Molecular Weight:
303.14
MDL number:
UNSPSC Code:
12352104

grade

battery grade

Quality Level

Assay

≥99.5% trace metals basis

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤5000 ppm (trace metals analysis)
≤5000 ppm Trace metals

mp

257.1-258.4 °C

application(s)

battery manufacturing

greener alternative category

SMILES string

FC(F)(F)S(=O)(=O)N([Na])S(=O)(=O)C(F)(F)F

InChI

1S/C2F6NO4S2.Na/c3-1(4,5)14(10,11)9-15(12,13)2(6,7)8;/q-1;+1

InChI key

YLKTWKVVQDCJFL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium bis(trifluoromethanesulfonyl)imide, abbreviated NaTFSI, is a white, crystalline salt. It is hygroscopic and deliquescent and usually stored under inert atmosphere. NaTFSI is soluble in water, carbonates (including dimethyl carbonate, diethyl carbonate, and propylene carbonate), acetonitrile, triglyme, dimethoxyethane, trimethyl phosphate, and many of the solvents commonly used in batteries.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

The primary application of NaTFSI is as an electrolytic salt in sodium-ion batteries. Researchers have found that NaTFSI dissolved in organic solvents[1] and, more recently, in ionic liquids[2,3] serve as high-performing electrolytes with a broad operating voltage exceeding 3 V. In addition, in the application of solid electrolytes for sodium-ion batteries, NaTFSI is often added to poly(ethylene oxide)-ceramic composites because it increases the ionic conductivity of the composite.[4,5]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service