Skip to Content
Merck
All Photos(1)

Key Documents

935956

Sigma-Aldrich

Sodium hexafluorophosphate solution

new
greener alternative

in ethylene carbonate and diethylcarbonate, 1.0 M NaPF6 in EC/DEC=50/50 (v/v), battery grade

Synonym(s):

1.0 M NaPF6 in EC/DEC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352600

grade

battery grade

Quality Level

form

solution

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

0.95-1.10 M (NaPF6)

impurities

≤100 ppm Acid
≤100.0 ppm H2O

cation traces

K: ≤10 ppm
Na: ≤10 ppm

application(s)

battery manufacturing

greener alternative category

General description

1.0 M NaPF6 in EC/DEC=50/50 (v/v) consists of a 1.0 M concentration of sodium hexafluorophosphate (NaPF6) dissolved in a 1:1 volume ratio of ethylene carbonate (EC) and diethyl carbonate (DEC). The product is a clear solution with strictly minimized impurities including low moisture, low trace metals, low chloride, and low sulfate.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

The main application of 1.0 M NaPF6 in EC/DEC=50/50 (v/v), battery grade is as an electrolyte in sodium-ion batteries (NIBs) Studies have found that when NaPF6 in a mixture of ethylene carbonate and diethyl carbonate (EC:DEC (1:1 v:v)) used as electrolyte in sodium-ion batteries (SIBs), there is an increase of the ionic conductivity, the cation transference number, and the binary diffusion coefficient thereby increasing the power density NaPF6 in EC/DEC can provide a stable and effective electrolyte solution that helps form a protective solid electrolyte interface (SEI) on the electrode surfaces, which reduces electrode breakdown and improves overall battery performance

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service