925438
MC-1
≥95%
Synonym(s):
10-(propylsulfonyl)decanoic acid, PSD-1000-M, Tricosaoxaheptacont-1-yl ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C60H120O27S
CAS Number:
Molecular Weight:
1305.64
UNSPSC Code:
12352103
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥95%
form
waxy solid
color
white
SMILES string
CCCS(CCCCCCCCCC(OCCOC)=O)(=O)=O
InChI key
CFSYKSGMBQEXLJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
MC-1 is an ampiphilic surfactant designed to enable a green method of peptide synthesis using water as the primary solvent in aqueous micellar catalysis. This product out-performs similar designer surfactants in enabling water based peptide synthesis, and does not require the usage of a co-solvent, affording a truly aqueous synthetic methodology. The structure of MC-1 is optimized that the inner micellar core simulates the properties of DMSO, a solvent ubiquitous with traditional peptide synthesis, while reducing the environmental drawbacks of using DMSO as the primary solvent.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service