Skip to Content
Merck
All Photos(1)

Key Documents

924199

Sigma-Aldrich

GelMA-RB

60% methacrylation, 1 umol Rhodamine B per gram material

Synonym(s):

GelMa-RB, Gelatin methacryloyl (GelMA)-Rhodamine B

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C32H47N8O11)n(C56H72N10O12)m
UNSPSC Code:
12352202
NACRES:
NA.23

Quality Level

form

powder

color

purple

storage temp.

2-8°C

Application

GelMA-Rhodamine B, 300 g Bloom is gelatin grafted with Rhodamine B that can be used in drug delivery research. GelMA is an engineered gelatin-based material with applications in tissue engineering, drug delivery, and 3D printing applications. Advantages of GelMa include biocompatibility, enzymatic cleavage, cell adhesion, and versatile mechanical properties. GelMa-Rhodamine B is uniquely suited for bioimaging and has several advantages − high fluorescence quantum yield, excellent sensitivity, and broad emission wavelength.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rhodamine Conjugated Gelatin Methacryloyl Nanoparticles for Stable Cell Imaging.
Xue Y, et al.
ACS applied bio materials, 3(10), 6908-6918 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service