Skip to Content
Merck
All Photos(1)

Key Documents

747378

Sigma-Aldrich

2,6-Dimethylaniline-15N

98 atom % 15N, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H1115N
Molecular Weight:
122.17
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

98 atom % 15N

Assay

98%

form

liquid

mass shift

M+1

SMILES string

CC1=CC=CC(C)=C1[15NH2]

InChI

1S/C8H11N/c1-6-4-3-5-7(2)8(6)9/h3-5H,9H2,1-2H3/i9+1

InChI key

UFFBMTHBGFGIHF-QBZHADDCSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service