Skip to Content
Merck
All Photos(1)

Key Documents

741159

Sigma-Aldrich

Phenylcyclohexane-13C12, d16

99 atom % 13C, 97 atom % D, 98% (CP)

Synonym(s):

Cyclohexylbenzene-13C12,d16

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13C6D513C6D11
Molecular Weight:
188.27
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

isotopic purity

99 atom % 13C
97 atom % D

Assay

98% (CP)

form

liquid

bp

239-240 °C (lit.)

mp

4-7 °C (lit.)

density

1.112 g/mL at 25 °C

mass shift

M+28

SMILES string

[2H][13c]1[13c]([2H])[13c]([2H])[13c]([13c]([2H])[13c]1[2H])[13C]2([2H])[13C]([2H])([2H])[13C]([2H])([2H])[13C]([2H])([2H])[13C]([2H])([2H])[13C]2([2H])[2H]

InChI

1S/C12H16/c1-3-7-11(8-4-1)12-9-5-2-6-10-12/h1,3-4,7-8,12H,2,5-6,9-10H2/i1+1D,2+1D2,3+1D,4+1D,5+1D2,6+1D2,7+1D,8+1D,9+1D2,10+1D2,11+1,12+1D

InChI key

IGARGHRYKHJQSM-NMPGWPKMSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids not in Storage Class 3

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service