Skip to Content
Merck
All Photos(1)

Key Documents

616516

Sigma-Aldrich

Bromoethane-2-d

98 atom % D

Synonym(s):

Ethyl bromide-2-d

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CDH2CH2Br
Molecular Weight:
109.97
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % D

Quality Level

bp

37-40 °C (lit.)

mp

-119 °C (lit.)

density

1.473 g/mL at 25 °C

mass shift

M+1

storage temp.

2-8°C

SMILES string

[2H]CCBr

InChI

1S/C2H5Br/c1-2-3/h2H2,1H3/i1D

InChI key

RDHPKYGYEGBMSE-MICDWDOJSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service