587974
Dimethylamine-d7
98 atom % D
Synonym(s):
Bis(trideuteriomethyl)deuterioamine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CD3)2ND
CAS Number:
Molecular Weight:
52.13
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12
Recommended Products
isotopic purity
98 atom % D
Quality Level
Assay
99% (CP)
form
liquid
bp
7 °C (lit.)
mass shift
M+7
SMILES string
[2H]N(C([2H])([2H])[2H])C([2H])([2H])[2H]
InChI
1S/C2H7N/c1-3-2/h3H,1-2H3/i1D3,2D3/hD
InChI key
ROSDSFDQCJNGOL-KBKLHOFPSA-N
Related Categories
Packaging
Standard 1 g quantity is packaged in a 25 mL Sure/Pac™ cylinder with stainless steel 1/4" male NPT valve. Nominal gas pressure at 21ºC is 13 psig for 1 g. This pressure is slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Recommended products
Stainless steel hose adapter Z14,683-8 or stainless steel body mini gas regulator Z51,354-7 is recommended.
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986
Legal Information
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 1 - Skin Corr. 1B - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
2A - Gases
WGK
WGK 1
Flash Point(F)
19.9 °F - closed cup
Flash Point(C)
-6.70 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service