Skip to Content
Merck
All Photos(1)

Key Documents

515914

Sigma-Aldrich

Bismuth(V) fluoride

99.9% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BiF5
CAS Number:
Molecular Weight:
303.97
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

99.9% trace metals basis

form

crystalline

reaction suitability

reagent type: catalyst
core: bismuth

density

5.40 g/mL (lit.)

SMILES string

F[Bi](F)(F)(F)F

InChI

1S/Bi.5FH/h;5*1H/q+5;;;;;/p-5

InChI key

BAHXPLXDFQOVHO-UHFFFAOYSA-I

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service