뒤로
Nunc
®
Universal Tubes in 2D bar coded latch rack
Products
제품 카테고리
tubes (3)
Manufacturer
용량(mL)
포장 단위
Technique
제품 필터
Showing 1-3 of 3
비교
제품 번호
설명
가격
TMO374501-BR
Nunc
®
Universal Tubes in 2D bar coded latch rack,
polypropylene, capacity 1.8 mL, flat bottom, external thread, untreated, case of 10 × 24 ea
확장
가격 보기
TMO374510-BR
Nunc
®
Universal Tubes in 2D bar coded latch rack,
polypropylene, capacity 2.0 mL, flat bottom, external thread, untreated, case of 10 × 24 ea
확장
가격 보기
TMO374500-BR
Nunc
®
Universal Tubes in 2D bar coded latch rack,
polypropylene, capacity (1.8 mL), flat bottom, external thread, untreated, case of 10 × 24 ea