주요 문서
G006159
GC Analysis of D & L-Carvone Enantiomers in Scotch Spearmint Essential Oil (Spiked with D-Carvone Enantiomer) on Astec® CHIRALDEX G-TA
suitable for GC
로그인조직 및 계약 가격 보기
About This Item
기술
gas chromatography (GC): suitable
검사 파라미터
column: CHIRALDEX G-TA, 30 m x 0.25 mm I.D., 0.12 μm (73033AST)
oven: 40 °C (1 min), then 2 °C/min to 170 °C (15 min)
inj. temp.: 250 °C
detector: FID, 250 °C
carrier gas: helium, 25cm/ sec set at 110°C
injection: 0.1 μL 100:1 split
liner: 4 mm I.D. cup injector
sample: Scotch Spearmint Oil + D- Carvone Spike, neat
응용 분야
flavors and fragrances
법적 정보
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)