주요 문서
57126
Supelco
SPE Tube Adapter
configured for AutoTrace Automated Systems, for use with 6mL tubes, pk of 6
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
12352200
포장
pk of 6
기술
solid phase extraction (SPE): suitable
응용 분야
food and beverages
호환성
configured for AutoTrace Automated Systems
for use with 6mL tubes
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Tube adapters serve many purposes. They can be used to stack one SPE tube on top of another to provide different selectivities. A larger empty syringe barrel can be stacked on top of a smaller SPE tube to act as a larger load reservoir. Or, they can serve as an adapter for positive pressure methods (e.g. from a syringe or air/ N2 line).
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)