23366
1 of 10
Supelco
20811
ORBO™ 80 Isocyanate Filter
Sigma-Aldrich
09772
Aluminum silicate
Millipore
M000037A0
Empty 37 mm Monitor Cassette with rings and plugs
520179
20812-U
ORBO™ 80 Coated Filter
742503
Calcium silicate
792640
Potassium silicate, anhydrous
643653
Aluminosilicate, mesostructured
03584
Kaolinite
854171
Vials, screw top, clear glass, 1.5 mL
pkg of 100 ea
100
environmentalindustrial hygiene
for use with 37mm Cassette
유사한 제품을 찾으십니까? 방문 제품 비교 안내
가장 최신 버전 중 하나를 선택하세요:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 문서 section.