주요 문서
Z757438
Sartorius Proline® Plus mechanical pippettor
single channel, volume 1-10 mL
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121500
NACRES:
NB.01
특징
electronic: no
single-channel
포장
pack of 1 ea
제조업체/상표
Sartorius 728090
용적
1-10 mL
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
The Sartorius Proline Plus pipettes of the Biohitfamily are high quality with a light pipetting force. With robust design, the Proline Plus is suitable for variable every day laboratory applications and covers the full range of 0.1μL to 10 mL.
  • Reduced pipetting forces prevent RSI and improve the results
  • Accurate and precise
  • Contamination free pipetting with Safe-Cone Filters
  • Robust design
  • Easiest pipette to maintain
법적 정보
Proline is a registered trademark of Sartorius Biohit Liquid Handling Oy
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)