주요 문서
Z742257
Adjustable Tube Rack Easel
Holds Rack up to 5.5 in. high and 8.5 in. wide, pack of 1 ea
동의어(들):
angled tube rack
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41122800
NACRES:
NB.35
material
acrylic
특징
adjustable 25, 35, 45 °
Holds Rack up to 5.5 in. high and 8.5 in. wide
포장
pack of 1 ea
제조업체/상표
Diversified Biotech TRAJ-1000
일반 설명
Universal, fixed position acrylic stand with neoprene pad for supporting tube racks. Holds tube-filled racks at a comfortable positing for easier pipetting.
특징 및 장점
  • Grooved support base for quick angle adjustment
  • Fully stabilized design
  • Pick an angle and start pipetting
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)