주요 문서
Z717304
Nichipet EX-Plus II pipette
volume 2-20 μL
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121500
NACRES:
NB.01
특징
electronic: no
single-channel
포장
pack of 1 ea
pkg of 1 ea
제조업체/상표
Nichiryo 00-NPL02-20
재고 정보
not available in (Not available in France)
용적
2-20 μL
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
The Nichipet EX-Plus ll is a new digital micropipette addition to the Nichiryo family of pipettors. The Nichipet EX-Plus ll offers a new, key feature of one-touch calibration.
Time and money spent on the calibration of pipettes is now greatly reduced!
  • UV resistant
  • Solvent resistant
  • Fully autoclavable (121°C for 20 minutes)
  • One-touch calibration
  • Ceramic pistons
  • Filtered nozzles
  • Easy, convenient locking lever
  • Ergonomic design
  • Single channel, manual, continuously adjustable volume
  • Advanced durability against organic solvent dispensing
  • PTFE (Fluorine resin) in the air tight chamber of the instrument
  • A Perfluoro rubber O-ring with low cubical expansion in the airtight parts
  • Plunger made from a material resistant to organic solvents
  • An alloy spring to avoid metal corrosion by organic solvents
  • Quick and easy to use
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)