주요 문서
Z708194
Pipettor guide
aligns pipetter with the multiwell plates, pkg of 1 ea
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121500
NACRES:
NB.01
포장
pkg of 1 ea
제조업체/상표
Diversified Biotech
일반 설명
Features a plastic sliding guide, above the multiwell plate to help align your pipetter with the multiwell plate wells. The sliding guide is clearly marked with column letters for fast and easy orientation. Compatible with multichannel pipetters; use with or without the 30° angled stand. A great tool to reduce eyestrain and tedious pipetting work.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)