주요 문서
Z659835
Bandelin SONOREX Ultrasonic bath accessories
positioning lid for Z659800 and Z659045
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41103706
NACRES:
NB.74
material
stainless steel
특징
for 4 beakers
포장
pack of 1 ea
제조업체/상표
Bandelin DE 514
재고 정보
not available in USA and Canada
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
When using chemically aggressive liquids, removing chemically aggressive or abrasive contamination or sonicating sample liquids, insert cups are used (indirect sonication). Because nothing may be placed on the bottom of the ultrasonic bath, use a suitable perforated lids/insert strips to hold the insert cups. Several insert cups can be sonicated with different liquids at the same time.
특징 및 장점
  • for holding insert cups
  • sound-absorbing
  • shortens the heating phase
법적 정보
SONOREX is a trademark of Bandelin Electronic GmbH & Co. KG
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)