주요 문서
HS23471A
Glove box holder, fits 1 glove box
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
42132201
NACRES:
NB.51
material
clear acrylic
특징
Holds 1 box
제조업체/상표
Heathrow Scientific HS23471A
크기
141x260x110 mm
일반 설명
The clear acrylic Glove Box Holder keeps gloves organized and ready for immediate use.
  • Clear acrylic holders include a polyester foam insert to securely hold a range of glove box sizes—snug fit ensures easy removal of gloves
  • All holders fit most major brands of boxes
  • Mounting screws included
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)