주요 문서
HS14521C
Aluminium weighing boat
D × H 70 mm × 12 mm , large
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121812
NACRES:
NB.35
제조업체/상표
Heathrow Scientific HS14521C
깊이 × 높이
70 mm × 12 mm , large
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
designed for general-purpose weighing, dispensing, storage, food processing, evaporating, or drying.
Boats also make good dust covers for open vessels.
  • All weigh boats are FDA/food-grade compliant
  • Disposable
  • Aluminum, Weighing Boats
Sturdy yet lightweight boats feature a smooth, flat bottom for sample handling and a flanged, crimped wall for rigidity. Finger tab handle can be etched for identification prior to weighing.
3 sizes
Packs of 100
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)