주요 문서
HS120196
Cardboard Slide Tray
Holds 20 x slides 76x26 mm, red (trim)
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41123302
NACRES:
NB.22
material
cardboard
red (trim)
특징
Holds 20 x slides 76x26 mm
포장
case of 72 trays
제조업체/상표
Heathrow Scientific HS120196
길이 × 너비 × 높이
208 × 341 × 9 mm
유사한 제품을 찾으십니까? 방문 제품 비교 안내
특징 및 장점
  • Easily organize slides with the index on the front cover that corresponds with the number grid inside
  • Recessed slide compartments and inside covers protect specimens. Slides cannot be touched when the cover is closed, reducing contamination
  • Front protection flaps are attached with durable tape for repeated use. Fold flaps flat under the slide holder to save space when in use
  • Thumb cut in slide compartments enables easy removal of slides with slight pressure at one end
  • Compartment holder is made from a double layer of densely pressed cardboard. Cover is a single layer cardboard
  • Disposable
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)