주요 문서
C079300
Sigma-Aldrich
Empty Synthesis Column
0.2μmol (Colourless Body)
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
12352200
NACRES:
NA.51
Quality Level
포장
pkg of 1 × 100 ea
호환성
configured for ABI
configured for PerkinElmer
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Empty Synthesis Columns are color coded and colorless empty column bodies with filters. C079300 empty synthesis column is a colorless body designed for ABI and Expedite Synthesizers.
법적 정보
ABI is a trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
Expedite is a trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)