주요 문서
BMSW110512500
Sigma-Aldrich
Accuris M1 Grade Calibration Weight Set
pack of 12 pieces, size 1-500 g, stainless steel
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41100000
NACRES:
NB.71
material
stainless steel
포장
pack of 12 pieces
제조업체/상표
Benchmark Scientific W1105-12500
크기
1-500 g
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Basic, compact balances with readability in the range of 0.1g to 0.01g can be calibrated with M1 Class weights.
Each individual weight and weight set from Accuris is supplied with an individual Certificate of Verification confirming that the weight(s) meet OIML regulations and tolerances for the specific weight class.
The economy M1 Calibration Weight Set includes 12 pieces: 1 x 1g, 2x2g, 1x5g, 1x10g, 2x20g, 1x50g, 1x100g, 2x200g, and 1x500g. A protective carrying case is included.
법적 정보
Accuris is a trademark of Benchmark Scientific, Inc.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)