주요 문서
BMSIPD960038781
Sigma-Aldrich
BeadBlaster 96 Ceramic grinding bar, 45° angled ends
bead diam. × L 3/8 × 7/8 , fits 50 mL tubes
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41100000
NACRES:
NA.82
material
ceramic
포장
pack of 100
제조업체/상표
Benchmark Scientific IPD960038781
bead diam. × L
3/8 × 7/8 , fits 50 mL tubes
호환성
BeadBlaster 96
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Ceramic grinding media is suitable for applications where highly efficient grinding must be paired with low grinding media abrasion. Non-ferrous grinding is absolutely essential in many processes, for instance in medical or pharmaceutical applications, in order to prevent undesired impurities in the grinding materials used. Ceramic grinding media are also suited for use in processes where mixing and dispersing effects play an important role.
특징 및 장점
  • low abrasion with high efficiency grinding
  • low impurity transfer
법적 정보
BeadBlaster is a trademark of Benchmark Scientific, Inc.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)