주요 문서
BMSIPD960025TC
Sigma-Aldrich
BeadBlaster 96 Grinding Jars for Very Hard Samples
tungsten carbide, capacity 25 mL, pack of 2
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41100000
NACRES:
NA.82
material
tungsten carbide
포장
pack of 2
제조업체/상표
Benchmark Scientific IPD9600-25TC
용량
25 mL
호환성
BeadBlaster 96
일반 설명
Very hard samples require a jar and grinding balls that are just as hard or harder for effective sample preparation. All of the jars feature threaded closures for leakproof performance with both solids and liquids.
Tungsten carbide is an exceptionally hard ceramic material. It is useful for grinding materials where the presence of trace metals may be a concern. Applications include preparation of samples for XRF and particle reduction of ores, minerals, metal oxides and cement clinkers. These jars are extremely durable and offer a long service life.
법적 정보
BeadBlaster is a trademark of Benchmark Scientific, Inc.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)