주요 문서
BAF370151000
CultureTemp® Warming Plate
AC/DC input 220 V AC, European 2-pin plug
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41102406
NACRES:
NB.76
material
anodized aluminum plate
포장
pack of 1 ea
제조업체/상표
Bel-Art F37015-1000
depth × W × H
25.4 × 24.9 × 5.7 cm
plate depth × W
17.8 × 20.3 cm
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Increase cell viability during cell passaging with this highly accurate warming plate that keeps cells at an optimal 37°C (+/-1°C ) outside of the incubator. The unit is water resistant and safe for cleaning with Ethanol or IPA.
특징 및 장점
  • Keep dishes outside the incubator longer; 37°C plate protects cell viability; no need to return to the incubator repeatedly
  • Reduces cell stress and allows enzymes like trypsin to work efficiently
  • Temperature remains uniform across the entire anodized aluminum warming plate t
  • Small footprint fits easily in a cell culture hood or on the workbench
  • UV resistant polypropylene housing will not break down under germicidal UV lights
  • 15-minute count down timer keeps track of time sensitive applications
법적 정보
CultureTemp is a registered trademark of SP Scienceware, Division of SP
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)