콘텐츠로 건너뛰기
Merck
모든 사진(11)

주요 문서

AMAB91903

Sigma-Aldrich

Monoclonal Anti-Msh6 Antibody Produced In Mouse

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, clone CL13844, affinity isolated antibody, buffered aqueous glycerol solution

동의어(들):

GTBP

로그인조직 및 계약 가격 보기


About This Item

UNSPSC 코드:
12352203

생물학적 소스

(Mouse)

Quality Level

결합

unconjugated

항체 형태

affinity isolated antibody

항체 생산 유형

primary antibodies

클론

CL13844, monoclonal

제품 라인

Prestige Antibodies® Powered by Atlas Antibodies

양식

buffered aqueous glycerol solution

정제법

(Protein A purified)

종 반응성

human

향상된 검증

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

농도

1.0mg/ml

기술

immunohistochemistry: 1:1000 - 1:2500
western blot: 1 μg/mL

동형

IgG2b

Ensembl | 인체 수납 번호

UniProt 수납 번호

배송 상태

wet ice

저장 온도

-10 to -25°C

유전자 정보

human ... MSH6(2956)

애플리케이션

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies™ protocols and other useful information.

특징 및 장점

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the

물리적 형태

Phospate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

기타 정보


Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user.

법적 정보

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

면책조항

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

적합한 제품을 찾을 수 없으신가요?  

당사의 제품 선택기 도구.을(를) 시도해 보세요.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable


가장 최신 버전 중 하나를 선택하세요:

시험 성적서(COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our 문서 section.

도움이 필요하시면 연락하세요. 고객 지원 부서

이 제품을 이미 가지고 계십니까?

문서 라이브러리에서 최근에 구매한 제품에 대한 문서를 찾아보세요.

문서 라이브러리 방문

자사의 과학자팀은 생명 과학, 재료 과학, 화학 합성, 크로마토그래피, 분석 및 기타 많은 영역을 포함한 모든 과학 분야에 경험이 있습니다..

고객지원팀으로 연락바랍니다.