주요 문서
20720
Hamilton® syringe, 1000 series GASTIGHT®, luer tip
1001LT, volume 1 mL, needle size (not included)
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41122001
eCl@ss:
32070500
material
glass body
니들 크기
(not included)
특징
without needle Luer tip
포장
pkg of 1 ea
제조업체/상표
Hamilton 81301
니들 길이
, n/a
용적
1 mL
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Hamilton® syringe is available in gas-tight type that minimizes the gas exchange in analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed. The tip is threaded in a luer lock syringe allowing the needle to be tightly screwed to the tip.
법적 정보
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)
문서 라이브러리 방문
Jeffrey S. Guy
Pharmacology for the Prehospital Professional, 54-54 (2009)
Don P. Wolf
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 192-192 (2012)