주요 문서
MACGLTVP1
Millipore
MAS-100 CG Ex® Validation Protocol
Letter format
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41116500
NACRES:
JA.11
Quality Level
호환성
for use with MAS-100 CG Ex® system
일반 설명
MAS-100 CG Ex® validation protocol is designed as a ready to use step by step guide to perform the validation activities for the compressed gas sampling pumps. The validation protocol in A4 format ensures regulatory compliance of the pumps and guarantees consistent performance.
애플리케이션
MAS-100 CG Ex® validation protocol is used for the validation of MAS-100 CG Ex® pumps which samples compressed gas for microbial contamination.
법적 정보
MAS-100 CG Ex is a registered trademark of MBV AG