콘텐츠로 건너뛰기
Merck
모든 사진(1)

주요 문서

70184-M

Sigma-Aldrich

Introductory pSTBlue-1 Perfectly Blunt® Cloning Kit - Novagen

로그인조직 및 계약 가격 보기


About This Item

UNSPSC 코드:
41106610
NACRES:
NA.54

제조업체/상표

Novagen®

Quality Level

저장 조건

OK to freeze
avoid repeated freeze/thaw cycles

배송 상태

dry ice

일반 설명

The Perfectly Blunt® Cloning Kits are designed for simplified cloning of DNA generated by PCR using any type of DNA polymerase. This approach enables the use of high-fidelity proofreading enzymes for amplification, thus decreasing the probability of generating mutations in the target sequence. In addition, under many conditions blunt cloning is more efficient than T-cloning, most likely due to the observation that the efficiency of single dA addition by Taq DNA polymerase varies significantly depending on the sequence context of the DNA ends, and even the number of PCR cycles performed (Novy 1996, Clark 1998, Brownstein 1996, Magnuson, 1996, Hu 1993).

With the Perfectly Blunt cloning protocol, you can go from PCR product to plating transformants in less than one hour with minimal hands-on time. The finished PCR product is converted to a blunt, phosphorylated form in a 15-minute reaction using premixed reagents. Following a 5-minute heat inactivation step, the treated insert is combined with the ready-to-use vector and ligated in an optimized 15-minute reaction. An exclusive 8-minute transformation procedure using highly efficient NovaBlue Singles Competent Cells (Cat. No. 70181) generates recombinant colonies that are easily visualized by blue/white screening.

Note that the Perfectly Blunt method is not limited to cloning PCR products; these kits are also suitable for cloning restriction fragments, cDNA, or sheared DNA with the same protocols.

Seven different vectors are available in Perfectly Blunt® Cloning Kits. Vector choices include those designed for general cloning, sequencing, optimal in vitro transcription/translation, and optimal protein expression in E. coli. Each vector is available in a kit containing sufficient reagents for 10, 20, or 40 reactions.

“Vector only” kits are also available in 20- and 40-reaction sizes without ligase and competent cells. For higher efficiency competent cells, also consider pSTBlue-1 Perfectly Blunt® Giga Cloning Kit (Cat. No. 71229).

The pSTBlue-1 vector is a multi-purpose cloning vector featuring a versatile multiple cloning region, blue/white screening, dual opposed T7/SP6 promoters and dual kanamycin/ampicillin resistance. Restriction sites producing 4-base 3′ overhangs are conveniently positioned near each end of the polylinker to facilitate the generation of unidirectional deletions using exonuclease III.

경고

Toxicity: Multiple Toxicity Values, refer to MSDS

기타 정보

Novy, R.E., et al. 1996. inNovations 6, 7. Clark, J.M. 1988. Nucleic Acids Res. 16, 9677. Brownstein, J.M., et al. 1996. BioTechniques 20, 1004. Magnuson, V.L., et al. 1996. BioTechniques 21, 700. Hu, G. 1993. DNA and Cell Biology 12, 763.

법적 정보

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Perfectly Blunt is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


시험 성적서(COA)

제품의 로트/배치 번호를 입력하여 시험 성적서(COA)을 검색하십시오. 로트 및 배치 번호는 제품 라벨에 있는 ‘로트’ 또는 ‘배치’라는 용어 뒤에서 찾을 수 있습니다.

이 제품을 이미 가지고 계십니까?

문서 라이브러리에서 최근에 구매한 제품에 대한 문서를 찾아보세요.

문서 라이브러리 방문

자사의 과학자팀은 생명 과학, 재료 과학, 화학 합성, 크로마토그래피, 분석 및 기타 많은 영역을 포함한 모든 과학 분야에 경험이 있습니다..

고객지원팀으로 연락바랍니다.