주요 문서
36-011-I
Sigma-Aldrich
Anti-nitro-α/α-Synuclein Antibody, clone nSyn12
clone nSyn12
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
12352203
클론
nSyn12, monoclonal
Quality Level
동형
IgG1κ
특이성
Clone nSyn12 is a mouse monoclonal antibody that detects nitrated human a and β-synuclein.
면역원
Full-length human recombinant Synuclein nitrated in vitro.
재구성
0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
저장 및 안정성
Stable for 1 year at +2°C to +8°C from date of receipt.
면책조항
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable