주요 문서
1.19133
Millipore
Mains charger
for use with MAS-100 NT®, with DC plug for serial numbers >110500
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41106200
NACRES:
NA.74
material
plastic hardware
Agency
non-compendial
특징
electronic
응용 분야
air monitoring
cosmetics
food and beverages
pharmaceutical
호환성
for use with MAS-100 NT®
일반 설명
Mains charger is an additional accessory for MAS-100 NT® air sampler. The MAS-100 NT® is the world′s best-selling air sampler, designed to monitor air microbiology in cleanrooms and hygienically ultra-sensitive areas. Its high recovery rate is based on the Andersen impaction principle. An integrated airflow sensor compensates for differences in pressure and temperature in the environment. The Mains charger for MAS-100 NT® has an input voltage of 100 to 240 V AC (47 to 63 Hz, 1.5 to 0.7 A) and output voltage of 24 V DC ±5% (2.7 A). The cable length varies depending upon the country.
애플리케이션
Mains charger is an additional accessory for MAS-100 NT® air sampler that is used for microbial monitoring of ambient air in controlled environments up to ISO 5 cleanrooms. Microbial air sampling devices are extensively used in Pharmaceutical and Personal care industries as well as Food, Beverage and Environmental applications.
법적 정보
MAS-100 NT is a registered trademark of MBV AG