주요 문서
1.17336
Millipore
MAS-100 Atmos® Filter Cover
for use with MAS-100 Atmos®
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41103403
NACRES:
NA.74
material
aluminum cover
설명
Compressed gas sampling
특징
autoclavable
제조업체/상표
MAS-100 Atmos®
응용 분야
compressed gas monitoring
cosmetics
food and beverages
pharmaceutical
호환성
for use with MAS-100 Atmos®
일반 설명
MAS-100 Atmos® Filter Cover is an accessory for MAS-100 Atmos® microbial compressed gas sampler′s HEPA filter (ref 1.17278).
The accessory is delivered along with 3 fitting screws.
The lower part of the sampling head of the MAS-100 Atmos® is fixed, as same as other mobile MAS-100® air samplers. Parts placed above are the instrument protection filter, filter holder, and plate holder. It is recommended not to remove these components before cleaning for routine applications. Disinfection can be done with clean tissue absorbed with 70% Ethanol or IPA.
애플리케이션
The MAS-100 Atmos® Filter Holder is used to provide proper placement to the HEPA Filter in MAS-100 Atmos® Microbial Compressed Gas Sampler.
법적 정보
MAS-100 is a registered trademark of MBV AG
MAS-100 Atmos is a registered trademark of MBV AG