주요 문서
1.17329
Millipore
MAS-100 Atmos® Battery Charger
for use with MAS-100 Atmos®
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41103403
NACRES:
NA.74
material
black polycarbonate adapter
설명
Compressed gas sampling
특징
electronic
제조업체/상표
MAS-100 Atmos®
응용 분야
compressed gas monitoring
food and beverages
pharmaceutical
호환성
for use with MAS-100 Atmos®
일반 설명
MAS-100 Atmos® Battery Charger is an accessory designed for recharging purpose.
The LED indicator of the power button displays a blue flashing light during charging and a permanent blue light when fully charged.
The power supply is delivered with a USB cable and country specific plugs.
애플리케이션
MAS-100 Atmos® Battery Charger is used to recharge the battery-operated MAS-100 Atmos® microbial compressed gas sampler.
기타 정보
Connect the mains charger to the USB type C port of the instrument to charge the battery. Charge the battery until the symbol on the local user interface indicates that the battery is full. The charging time is approx. 7.5 hours.
법적 정보
MAS-100 Atmos is a registered trademark of MBV AG