주요 문서
Z677477
Camatril Velours nitrile flock lined gloves
size XXL
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
46181504
NACRES:
NB.35
material
green gloves
green nitrile
크기
XXL
무균
non-sterile
특징
CE compliant
disposable
latex free
powder-free
reusable
standard type EN 374 (cat. III)
standard type EN 388 (cat. III)
포장
package of 100 ea
제조업체/상표
KCL 730 - 11
KCL 730
길이 × thickness
300 mm × 0.4 mm
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Camatril Velours green nitrile flock lined gloves are manufactured from high quality nitrile ensuring effective chemical resistance for use in numerous fields. The remarkably high abrasion, cut and puncture resistance allows work with sharp edged materials.
Good flexibility, anatomical design and excellent fit ensure a good sense of touch. The flock lining offers additional wearing comfort.
  • Ergonomic fit ensures good sensitivity
  • Roughened palm for excellent grip when wet
  • High mechanical resistance
  • Cotton flock lining improves comfort
  • Free from allergenic substances such as thiurames, benzothiazoles and thiourea
  • CE compliant
법적 정보
Camatril is a trademark of KCL GmbH
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)