주요 문서
Z677418
Dermatril P nitrile gloves
size XL
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
46181504
NACRES:
NB.35
material
blue gloves
blue nitrile
크기
XL
무균
non-sterile
특징
CE compliant
disposable
latex free
powder-free
포장
pack of 50 ea (boxes of 50)
제조업체/상표
KCL 743 - 10
KCL 743
길이 × thickness
270 mm × 0.2 mm
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Dermatril P is an extended cuff, blue nitrile, powder-free glove providing a high level of protection against chemical and bacteriological contamination. The Dermatril P disposable glove also displays very good physical durability in comparison to conventional disposable gloves.
All Dermatril P disposable gloves are INDIVIDUALLY marked with the appropriate standards mark applied on the inside using a food-friendly dye.
특징 및 장점
  • Anti-static
  • Free from allergenic substances such as thiurames, benzothiazoles and thiourea
  • Roughened fingertips for better grip
  • Rolled cuff edge for easier donning
  • CE compliant
  • Meet standards: EN 374 (cat. III), EN 388 (cat. II), EN 61340-5-1, ASTM 1671:2001
법적 정보
Dermatril is a trademark of KCL GmbH
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)