주요 문서
Z550833
Spilfyter® hazmat spill control, polypropylene
Pad, W × L 16 in. × 18 in., double weight
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
47131905
NACRES:
NB.35
포장
pkg of 100 ea
너비 × 길이
16 in. × 18 in.
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Spilfyter® products are made from a specially formulated, lint-free, 100% PP fabric that is virtually inert and will absorb any liquid with which they come in contact. They are able to collect up to 10 times their own weight in waste material (depending upon its viscosity) in 30s. Saturated Spilfyter® products can be landfilled, neutralized, or incinerated in accordance with federal, state and local regulations.
법적 정보
Spilfyter is a registered trademark of National Packaging Services Corp.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)