주요 문서
Z260258
FORTUNA® Optifix® Safety bottle-top dispenser
Safety model, volume 20-100 mL
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121500
NACRES:
NB.01
특징
subdivision 2.0 mL
with recirculating valve
제조업체/상표
Fortuna Graf 101.107-51
부피 측정 범위
20-100 mL
용적
20-100 mL
피팅
GL 40
GL 45
유사한 제품을 찾으십니까? 방문 제품 비교 안내
관련 카테고리
일반 설명
For dispensing strong acids and aggressive chemicals
Features:
  • PTFE encased dispensing piston with sealing lip at its end
  • FIX adapter made of PTFE
  • Valve block made of PTFE, no metal springs in the valve - no corrosion
  • Visible dispensing process by the use of a dispensing cylinder made of glass, coated with plastic-protection against breakage
  • Ergonmic finger protection cap
  • Fast and easy volume setting with additional fine adjustment
Supplied as Optifix® Safety dispenser, 2 PTFE adapters GL40 + GL45 & 1 PTFE intake tube
법적 정보
FORTUNA is a registered trademark of Poulten & Graf Ltd.
Optifix is a registered trademark of Poulten & Graf Ltd.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)