주요 문서
SLW1517/08
Pyrex® Media-Lab Bottles, with-out cap and pouring ring, with printed trace code
capacity 500 mL
동의어(들):
Pyrex, SciLabware 1517/08
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121800
NACRES:
NB.17
material
borosilicate glass bottle
round bottle
white graduation
조인트
Joints threaded neck (GL45)
무균
non-sterile
특징
graduations
without cap
with printed trace code
without pouring ring
포장
pack of 10 ea
제조업체/상표
SciLabware 1517/08
용량
500 mL
직경 × 높이
86 mm × 176 mm
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
  • Bottles only. Supplied without cap or pouring ring
  • Complies with ISO 4796
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download from www.scilabware.com
  • Sloping shoulders for smooth pouring action
  • White graduations and marking spot
  • Manufactured from temperature and chemically resistant borosilicate glass
특징 및 장점
  • Bottles only. Supplied without cap or pouring ring.
  • Complies with ISO 4796.
  • Printed with Trace Code for downloadable batch certificate.
  • Sloping shoulders for smooth pouring action.
  • White graduations and marking spot.
  • Manufactured from temperature and chemically resistant borosilicate glass.
  • Choice of caps and pouring rings available separately.
  • The 25 ml size bottle features a glass lip and does not require a pouring ring.
법적 정보
PYREX is a registered trademark of Corning, Inc.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)