주요 문서
PL800GA-HYP-8
Plas-Labs replacement Hypalon® gloves
for 2 piece glove set, size 8 (for 8 in. ports)
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
46181504
NACRES:
NB.35
material
Hypalon®
크기
8 (for 8 in. ports)
특징
reusable
for use with glove box sleeves
포장
pack of 1 pair
제조업체/상표
Plas-Labs 800GA-HYP-8
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Plas-Labs glovebox gloves are available as either one-piece or two-piece assemblies with interchangeable sleeves and gloves. This enables personnel to use the same glove box by changing the size or type of glove being used. Choose from four rubber types. Gloves are 15 mil thickness unless noted otherwise.
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
  • White Hypalon® rubber for resistance to abrasion, oxidizing chemicals, ozone and UV light
  • Black neoprene rubber for chemical resistance
  • Black butyl rubber for highest impermeability to water vapor, gases, and toxic fluids
  • Green nitrile rubber for resistance to oil, fuel, and acids
  • Latex rubber for flexibility
법적 정보
Hypalon is a registered trademark of E. I. du Pont de Nemours and Company
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)