주요 문서
NORTCM400
Bruker tapered PTFE caps for Match tubes
for NORS405001
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121800
NACRES:
SB.32
특징
for NORS405001
포장
pack of 5 ea
제조업체/상표
Norell TCM400
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Bruker Match tube caps are specially machined from PTFE fluoropolymer, are color coded by tube size, and correspond to the color of the tube clamp in the Match Insert Assembly, This enables quick identification of matching components. The tube cap incorporates a slightly tapered inner diameter. Initially, the cap aligns and pushes easily onto the tube, it then becomes progressively tighter as the tube reaches full depth within the cap, creating a positive seal to preserve volatile sample solutions throughout short or long term storage.
법적 정보
Match is a trademark of Bruker Biospin Corp.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)