주요 문서
ESYNTH013
Sigma-Aldrich
SynLectro Lead Electrode
size 3 mm × 20 mm × 60 mm
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
81141502
NACRES:
NB.61
크기
3 mm × 20 mm × 60 mm
일반 설명
SynLectro Lead Electrode is a 3x20x60 mm electrode designed for use with the SynLectro Electrolysis Platform for synthetic electrochemistry.
SynLectro Electrolysis Platform
The SynLectro Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component, designed with consistency, reproducibility, and safety in mind.
법적 정보
SynLectro is a trademark of Merck KGaA, Darmstadt, Germany
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)