주요 문서
ESYNTH004
Sigma-Aldrich
SynLectro 50 mL Jacketed Glass Cell with PTFE stopper
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
81141502
NACRES:
NB.61
일반 설명
SynLectro 50 mL Jacketed Glass Cell with PTFE Stopper is the part of the SynLectro Electrolysis Platform for synthetic electrochemistry.
Features and Benefits:
  • Designed to simplify setup, optimize reactivity, and improve safety
  • Side-arm enables connection to reflux condenser or inert gas
  • Jacket supports thermal control
  • Chemical resistant material (PTFE)
  • Protects electrical contacts from electrolytes
  • Sleeve (optional) prevents seized joints
SynLectro Electrolysis Platform
The SynLectro Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component, designed with consistency, reproducibility, and safety in mind.
법적 정보
SynLectro is a trademark of Merck KGaA, Darmstadt, Germany
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)