주요 문서
BR782155
BRAND® lids for microplates BRAND®plates
plate size 96 wells, without condensation rings
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121800
NACRES:
NB.43
특징
without condensation rings
제조업체/상표
BRAND 782155
플레이트 크기
96 wells
유사한 제품을 찾으십니까? 방문 제품 비교 안내
관련 카테고리
일반 설명
To protect samples, reaction batches or cell cultures from both contamination and evaporation, compatible lids are available for all BRANDplates® microplates. The lids are made of crystal-clear polystyrene. They are optimally matched to the corresponding BRANDplates® microplates, so that plates with lids can also be stacked. The cut-corner sets the orientation of the lid. For 96-well standard plates, lids with condensation rings are available.
법적 정보
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)