주요 문서
BR4600101
BRAND® Dispensette® S Analog-adjustable bottle-top dispenser
volume range 0.1-1 mL, with recirculation valve
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121500
NACRES:
NB.01
특징
incremental 0.02 mL
DE-M marking
with recirculation valve
제조업체/상표
BRAND 4600101
용적 범위
0.1-1 mL
피팅
(GL 24-25)
(GL 28/S 28)
GL 32
GL 38
S 40
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Dispensette S is suitable for an amazingly broad range of applications, particularly for many acids, saline solutions and organic solvents.
특징 및 장점
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
기타 정보
Analog-adjustable - Rapid and safe volume setting with new internal scalloped track. Simple calibration adjustment with a supplied tool.
법적 정보
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)
문서 라이브러리 방문
프로토콜
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
관련 콘텐츠
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.