주요 문서
BAH942404003
Scienceware® Lab Companion perforated shelf
fits 23 L, 35 L, 45 L cabinet vacuum desiccators, pack of 1 ea
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41104510
NACRES:
NB.43
포장
pack of 1 ea
제조업체/상표
Bel-Art H94240-4003
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
SP Bel-Art Perforated Shelf, available as a replacement of or addition to Lab Companion™ cabinet style vacuum desiccators. Perforated shelves permit efficient airflow throughout cabinet; all models allow for additional shelves to be added.
법적 정보
Lab Companion is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)